missing translation for 'onlineSavingsMsg'
Learn More

HEPES, free acid, >98%, MP Biomedicals™

£37.75 - £1895.00

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394379
View Documents
MP Biomedicals
0210192625
25 g
£37.75
25g
Please to purchase this item. Need a web account? Register with us today!
11304389
View Documents
MP Biomedicals
0210192650
50 g
£60.85
50g
Please to purchase this item. Need a web account? Register with us today!
11314389
View Documents
MP Biomedicals
0210192680
100 g
£86.85
100g
Please to purchase this item. Need a web account? Register with us today!
11324389
View Documents
MP Biomedicals
0210192683
250 g
£196.00
250g
Please to purchase this item. Need a web account? Register with us today!
11334389
View Documents
MP Biomedicals
0210192690
500 g
£342.00
500g
Please to purchase this item. Need a web account? Register with us today!
11344389
View Documents
MP Biomedicals
0210192691
1 kg
£533.00
1kg
Please to purchase this item. Need a web account? Register with us today!
11354389
View Documents
MP Biomedicals
0210192694
5 kg
£1895.00
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
CHEBI:42334

Specifications

7365-45-9
>98%
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
238.3
HEPES
White
C8H18N2O4S
hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 230-907-9

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.