missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free

£115.00 - £740.00

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10723894
View Documents Promotion Details
Thermo Scientific Chemicals
329850500
50 g
£115.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10585681
View Documents Promotion Details
Thermo Scientific Chemicals
329855000
500 g
£740.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
C8H18N2O4S
MFCD00006158
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
CHEBI:42334
OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

(0.5 M in water), (0.5 M in water) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm
7365-45-9
White
Crystalline Powder
Authentic
C8H18N2O4S
Plastic bottle
15, 4689
Solubility in water: 704g/L (20°C).
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
≥99%
HEPES free acid
(0.5M in water)
234.0°C to 238.0°C
5 to 6.5 (0.5 M aq. soln.)
99+%
0.5% max. (105°C)
MFCD00006158
23, V,2, 379
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
23831
238.3
Molecular Biology
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

missing translation for 'einecsNumber' : 230-907-9

missing translation for 'rtecsNumber' : TL6809000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only