missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HEPES Lysis Buffer with NP-40, 2X
£44.70 - £63.50
Specifications
Content And Storage | Ambient temperatures |
---|---|
Description | Clear |
Format | Liquid |
Product Type | Cell Lysis Reagent |
Concentration | 2X |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466019
|
Thermo Scientific™
J63867.=E |
125mL |
£44.70
125mL |
In Stock
Log in to see stock. |
|||||
15476019
|
Thermo Scientific™
J63867.AK |
250mL |
£63.50
250mL |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ambient temperatures | |
Liquid | |
2X |
Clear | |
Cell Lysis Reagent |