missing translation for 'onlineSavingsMsg'
Learn More

HEPES protein extraction buffer (5X), Thermo Scientific™

£49.05 - £50.45

Specifications

Chemical Name or Material HEPES protein extraction buffer
Color Colorless
Concentration 5X
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15425569
View Documents
Thermo Scientific Alfa Aesar
J63297.AK
250 mL
£50.45
250mL
Please to purchase this item. Need a web account? Register with us today!
15435569
View Documents
Thermo Scientific Alfa Aesar
J63297.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

HEPES protein extraction buffer
Colorless
5X
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications