missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals HEPES sodium salt, 99% Product Code.: 10532554

Thermo Scientific Chemicals HEPES sodium salt, 99%

Product Code. 10532554
1 kg
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10532554

Brand: Thermo Scientific Chemicals 215000010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm
Absorbance Conditions (0.1M aq. soln.)
Color White
pH 9.5 to 10.5 (1% aq. soln.)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99%
Infrared Spectrum Authentic
Formula Weight 260.27
Percent Purity ≥98.5% (on anhydrous substance)
Chemical Name or Material HEPES sodium salt, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455462 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number215000010,215001000,215000250,215005000
Lot NumberA0401478 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number215000010,215001000,215000250,215005000
Lot NumberA0447794 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number215000010,215001000,215000250,215005000
Lot NumberA0443576 Certificate TypeCertificate of Analysis Date28/06/2022 Catalog Number215000010,215001000,215000250,215005000
Lot NumberA0407573 Certificate TypeCertificate of Analysis Date31/10/2021 Catalog Number215000010,215001000,215000250,215005000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals HEPES sodium salt, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.