missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

£112.56 - £367.83

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10328103
View Documents Promotion Details
Thermo Scientific Chemicals
327261000
100 g Plastic bottle
£112.56
100g
Please to purchase this item. Need a web account? Register with us today!
10653332
View Documents Promotion Details
Thermo Scientific Chemicals
327265000
500 g Plastic bottle
£367.83
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

75277-39-3
260.28
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
C8H17N2NaO4S
MFCD00036463
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
CHEBI:46758
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

(0.1 M in H2O), (0.1 M in H2O) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
75277-39-3
White
Crystalline Powder
Authentic
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
2724248
260.27
Molecular Biology
(0.1M in H2O)
248.0°C
5.0 to 15.0 (1% soln.)
99+%
C8H17N2NaO4S
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
260.28
CHEBI:46758
≥99%
HEPES sodium salt, 99+%
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only