missing translation for 'onlineSavingsMsg'
Learn More

HEPPSO, 0.2M buffer soln., pH 7.0, Thermo Scientific™

Product Code. 15416039
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-78-0
Molecular Formula C9H20N2O5S
Molecular Weight (g/mol) 268.328
MDL Number MFCD00038351
InChI Key GIZQLVPDAOBAFN-UHFFFAOYSA-N
PubChem CID 100205
ChEBI CHEBI:32951
IUPAC Name 2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCO)CC(CS(=O)(=O)O)O

Specifications

Color Colorless
Concentration 0.2 M
pH 7.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material HEPPSO buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'