missing translation for 'onlineSavingsMsg'
Learn More

Heptafluoro-2-iodopropane, 97%, stab. with copper powder, Thermo Scientific Chemicals

£29.00 - £226.00

Chemical Identifiers

CAS 677-69-0
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.926
MDL Number MFCD00042089
InChI Key BBZVTTKMXRPMHZ-UHFFFAOYSA-N
Synonym heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
PubChem CID 33978
IUPAC Name 1,1,1,2,3,3,3-heptafluoro-2-iodopropane
SMILES C(C(F)(F)F)(C(F)(F)F)(F)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379215
View Documents
Thermo Scientific Alfa Aesar
L16523.06
5 g
£29.00
5g
Please to purchase this item. Need a web account? Register with us today!
11389215
View Documents
Thermo Scientific Alfa Aesar
L16523.14
25 g
£77.00
25g
Please to purchase this item. Need a web account? Register with us today!
11399215
View Documents
Thermo Scientific Alfa Aesar
L16523.22
100 g
£226.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

677-69-0
295.926
BBZVTTKMXRPMHZ-UHFFFAOYSA-N
33978
C(C(F)(F)F)(C(F)(F)F)(F)I
C3F7I
MFCD00042089
heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
1,1,1,2,3,3,3-heptafluoro-2-iodopropane

Specifications

677-69-0
2.1
C3F7I
1.33
1841228
heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
C(C(F)(F)F)(C(F)(F)F)(F)I
295.926
295.93
Heptafluoro-2-iodopropane, stabilized with copper powder
-56°C
38°C
(CF3)2CFI
MFCD00042089
Light sensitive
BBZVTTKMXRPMHZ-UHFFFAOYSA-N
1,1,1,2,3,3,3-heptafluoro-2-iodopropane
33978
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

missing translation for 'einecsNumber' : 211-643-3

missing translation for 'rtecsNumber' : TZ3925000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only