missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, 99%, for protein sequence analysis, Thermo Scientific Chemicals

£596.00 - £888.00

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
13488558
View Documents
Thermo Scientific Acros
411221000
100 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10674012
View Documents
Thermo Scientific
411221500
150 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

375-22-4
214.04
YPJUNDFVDDCYIH-UHFFFAOYSA-N
9777
2,2,3,3,4,4,4-heptafluorobutanoic acid
C4HF7O2
MFCD00004171
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
CHEBI:39426
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

375-22-4
-17.5°C
1.6400g/mL
Authentic
C4HF7O2
1.3
3265
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
YPJUNDFVDDCYIH-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutanoic acid
9777
214.04
Powder
(0.1% aqueous solution), (0.1% aqueous solution) at 0nm, 0.004 max. at 280nm, 0.006 max. at 254nm, 0.15 max. at 230nm
Beige to Brown-Orange
120.0°C (755.0 mmHg)
99%
CF3CF2CF2CO2H
MFCD00004171
1.64
Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
214.04
CHEBI:39426
99%
Heptafluorobutyric Acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-786-3

missing translation for 'rtecsNumber' : ET4025000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only