missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals

£32.76 - £316.58

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10771831
View Documents Promotion Details
Thermo Scientific Acros
172800250
25 g Glass bottle
£32.76
25g
Please to purchase this item. Need a web account? Register with us today!
10772021
View Documents Promotion Details
Thermo Scientific Acros
172801000
100 g Glass bottle
£90.75
100g
Please to purchase this item. Need a web account? Register with us today!
10639263
View Documents Promotion Details
Thermo Scientific Acros
172805000
500 g Glass bottle
£316.58
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

375-22-4
214.04
YPJUNDFVDDCYIH-UHFFFAOYSA-N
9777
2,2,3,3,4,4,4-heptafluorobutanoic acid
C4HF7O2
MFCD00004171
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
CHEBI:39426
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

375-22-4
Colorless to Yellow
120.0°C (755.0 mmHg)
99%
CF3CF2CF2CO2H
MFCD00004171
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
YPJUNDFVDDCYIH-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutanoic acid
9777
214.04
Liquid
-17.5°C
1.6400g/mL
Authentic
C4HF7O2
1.3
1.64
Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
214.04
CHEBI:39426
99%
Heptafluorobutyric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-786-3

Product Identifier
  • Heptafluorobutyric acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only