missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, For pesticide residue analysis, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
£72.77 - £557.74
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15654220
|
Honeywell-Chromasolv
34495-1L |
1 L |
£72.77
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664220
|
Honeywell-Chromasolv
34495-2.5L |
2.5 L |
£160.71
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15674220
|
Honeywell-Chromasolv
34495-4X2.5L |
4 x 2.5 L |
£557.74
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
142-82-5 | |
0.07 | |
433°F | |
CH3(CH2)5CH3 | |
83 mmHg (37.7°C), 40 mmHg (20°C) | |
1730763 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2g/mol | |
CHROMASOLV™ |
0.684g/mL at 25°C | |
−4°C (24.8°F) | |
C7H16 | |
MFCD00009544 | |
UN1206 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
≥99% (GC) | |
Heptane |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
missing translation for 'rtecsNumber' : MI7700000