missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane CHROMASOLV™, for HPLC, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
100 mL
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
Melting Point | -91°C |
Density | 0.684g/mL at 25°C |
Boiling Point | 98°C |
Quantity | 6 x 1 L |
Refractive Index | n20/D 1.387 |
Linear Formula | CH3(CH2)5CH3 |
Vapor Pressure | 83mmHg (37.7°C) |
Beilstein | 1730763 |
Formula Weight | 100.2g/mol |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Product Identifier
- Heptane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'