missing translation for 'onlineSavingsMsg'
Learn More

Heptane, Extra Pure, Fraction, SLR, Fisher Chemical™

Product Code. 10112050
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Heptane, Extra Pure, Fraction, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00081849
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
PubChem CID 8900
ChEBI CHEBI:43098
SMILES CCCCCCC

Specifications

Packaging Amber glass bottle
Quantity 500 mL
Color Colorless
IUPAC Name heptane
Formula Weight 100.2g/mol
Vapor Pressure 48mbar at 20°C
Viscosity 0.4 mPaS at 20°C
Vapor Density 3.5
Melting Point -90°C
Boiling Point 90°C to 100°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'