missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptanes, mixture of isomers, Reagent Grade, contains ≥25% n-heptane, Honeywell
Click to view available options
Quantity:
4 L
Specifications
CAS | 142-82-5 |
Density | 0.693 g/mL at 25 °C |
Boiling Point | 92°C to 100°C |
Flash Point | 8°C |
Packaging | Poly Bottle |
Molecular Formula | C7H16 |
Linear Formula | C7H16 |
MDL Number | MFCD00009544 |
Quantity | 4 L |
UN Number | UN1206 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'