missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic acid, 98%, Thermo Scientific Chemicals

£23.01 - £144.72

Chemical Identifiers

CAS 111-14-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00004426
InChI Key MNWFXJYAOYHMED-UHFFFAOYSA-N
Synonym enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
PubChem CID 8094
ChEBI CHEBI:45571
IUPAC Name heptanoic acid
SMILES CCCCCCC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10783351
View Documents Promotion Details
Thermo Scientific Acros
164172500
250 mL Glass bottle
£23.01
250mL
Please to purchase this item. Need a web account? Register with us today!
10092000
View Documents
Thermo Scientific Acros
164170010
1 L Glass bottle
£55.88
1 L
Please to purchase this item. Need a web account? Register with us today!
13468398
View Documents
Thermo Scientific Acros
164170025
2.5 L Glass bottle
£144.72
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-14-8
130.19
MNWFXJYAOYHMED-UHFFFAOYSA-N
8094
heptanoic acid
C7H14O2
MFCD00004426
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
CHEBI:45571
CCCCCCC(=O)O

Specifications

111-14-8
White
223.0°C to 223.4°C
Authentic
C7H14O2
1.4210 to 1.4240
02,338
0.91
Solubility in water: 0.24g/100mL (15°C). Other solubilities: soluble in alcohol, ether, dmf, chloroform, petrol, soluble in most organic solvents
CCCCCCC(=O)O
130.19
CHEBI:45571
130.19
Powder
-10.5°C
0.9100g/mL
>112°C
98%
CH3(CH2)5CO2H
MFCD00004426
15,4695
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
MNWFXJYAOYHMED-UHFFFAOYSA-N
heptanoic acid
8094
3.40 mPa.s (30°C)
98%
Heptanoic acid, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-838-7

missing translation for 'rtecsNumber' : MJ1575000

missing translation for 'tsca' : TSCA

Product Identifier
  • Heptanoic acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only