missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 10502191
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2528-61-2
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD00000763
InChI Key UCVODTZQZHMTPN-UHFFFAOYSA-N
Synonym enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
PubChem CID 17313
IUPAC Name heptanoyl chloride
SMILES CCCCCCC(Cl)=O

Specifications

Melting Point -84°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 173°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5COCl
Refractive Index 1.4295 to 1.4315
Quantity 100 g
Beilstein 02, 340
Specific Gravity 0.96
Solubility Information Solubility in water: hydrolysis. Other solubilities: slightly soluble in carbontetrachloride,soluble in ethylether,ligroin
Formula Weight 148.63
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Heptanoyl chloride, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'