Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Brand: Thermo Scientific™ 51901071
406.61 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 51
Additional Details : Weight : 1.00000kg
Description
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95 | |
Left Hand Glass with Metal Sheet and 3 Media Valve Holes | |
Side Window Modifications | |
Factory Installed |