missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm Incubation Chamber 750L
GREENER_CHOICE

Product Code. 18774584 Shop All Thermo Scientific Products
Click to view available options
Door Style:
Glass
Solid (outer), Glass (inner)
Unit Size:
1 unit
This item is not returnable. View return policy

Product Code. 18774584

Brand: Thermo Scientific™ EIG750

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.

Produces precise temperature conditions above and below ambient, ideally suited for microbiological incubation, and heated or cooled incubated storage.

  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu. ft.
Capacity (English) Shelf 66.1 lbs
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Ethernet
Capacity (Metric) 750 L
Chamber Material Powder-coated Cold Rolled Steel
Lighting No
Material Powder-coated Cold Rolled Steel
No. of Shelves 3
Plug Type Nema 5-20P
Temperature Range (Metric) 0 to 70°C
Type Environmental Chamber
Display 7 in. touchscreen user interface
Door Style Glass
No. of Casters 4
Ports 2 access ports
Programmability Temperature ramping
Shelf Area (English) 4.9 ft²
Shelf Area (Metric) 0.46 m²
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor - programmable
Temperature Resolution 0.1°C
Temperature Stability ≤ ±0.1°C @ 20°C to 37°C
Temperature Uniformity ≤ ±0.5°C @ 20°C to 37°C
Warranty 24 months
Width (English) 42.52 in.
Width (English) Interior 33.07 in.
Width (English) Shelf 22.6 in.
Width (Metric) 1080 mm
Width (Metric) Interior 840 mm
Width (Metric) Shelf 795 mm
Height (English) 72.24 in.
Height (English) Interior 57.09 in
Height (Metric) 1835 mm
Height (Metric) Interior 1450 mm
Depth (English) 34.84 in.
Depth (English) Interior 24.8 in.
Depth (Metric) 885 mm
Depth (Metric) Interior 630 mm
Shipping Weight (English) 820 lbs
Shipping Weight (Metric) 372 kg
Electrical Requirements 100/240V, 50/60 Hz, 1400W
Voltage 100/115/220/230 V
Wattage 900 W
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.