Learn More
Thermo Scientific™ Heratherm™ Refrigerated Incubators
Offers precise temperature control, increased energy efficiency, and ease of usability. Equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results.
Brand: Thermo Scientific™ 51031567
8813.61 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
View more versions of this product
Description
Achieve untapped potential in incubation with the temperature precision you need for applications that demand it. With a temperature range of +5 to +70°C, Thermo Scientific™ Heratherm™ Refrigerated Incubators are equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results. With no hazardous refrigerants, these units use less energy than the conventional product, making it the ideal choice for energy-conscious labs.
- Temperature range 5- 70°C
- Available in 2 sizes: benchtop model 178L / 6.3 cu.ft. and a floormodel with casters 381L / 13.5 cu.ft.
- Great temperature uniformity and stability to keep the samples in a safe environment –> uniformity as low as +/-0.3°C (20°C), stability as low as +/-0.1°C
- Energy consumption as low as 80Wh/h(benchtop) / 150Wh/h (floorstanding) at 20°C - Note that comparable compressor cooled units consume up to 6 times this amount*
- CFC free / HFC free polyurethane insulation foam for outstanding temperature performance and condensation prevention
- Heated door with intelligent energy control to ensure no condensation – even at low temperatures
- Sophisticated timer function with weekly,daily or real time mode
- Intuitive programmability function for defined temperature ramping, soak/cool and dwell: store up to 10 programs with 10 easysteps
- Adjustable over temperature shut-off for protocols that require setting of over temperature for protection – (class 2 according to norm DIN12880)
- Safe viewing of samples through internal glass door with limited impact on temperature
- Safe containment with automatic over and under temperature alarm
- Standard access port for data monitoring (large diameter of 42mm / 1.65 in.)
- Flexible shelf system for optimal loading, easy to remove for cleaning, with non-tip protection
- Stainless steel interior (1.4301 / ASTM 304) and rounded corners for easy cleaning
*Based on comparison with models BK6160/Precision815
Note: stated values are based on norm DIN12880 testing
Services
2 years

Specifications
13.5 cu. ft. | |
Heratherm refrigerated incubator IMP400, w/ electrical outlet, 230V, 50/60Hz | |
Stainless Steel (304/1.4301) | |
Coated Steel | |
EU plug CEE7/7 | |
Automated Incubator | |
Solid outer door & internal glass door | |
Internal Electrical Outlet: EU (CEE 7/4 plug and interior outlet) | |
IMP400 | |
1.6 in. (42mm) | |
0.1°C | |
524 x 544 x 1335 mm (20.6 x 21.4 x 52.6 in.) | |
60.8 in. (casters not included: 7.1 in.) | |
476 lb. | |
368 lb. | |
230 V, 50/60 Hz, 6.0 A, 1360 w |
CE | |
381 L | |
20.79 x 21.06 in. (528 x 498 mm) | |
2/17 | |
5°C to 70°C | |
Large Vacuum Fluorescent | |
Energy efficient | |
CFC free/HFC free polyurethane insulation foam | |
1 | |
Peltier technology: no refrigerant | |
≤0.3°C at 20°C; ≤0.7°C at 37°C | |
770 x 778 x 1545 mm ( 30.3 x 30.6 x 60.8 in.) | |
1545 mm (casters not included: 180 mm) | |
216 kg | |
167 kg | |
230 V |
Special Offers
- Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.