missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Heratherm Stability Chamber 750 L
Gain confidence in your product when testing stability and shelf life with robust Heratherm Stability chambers designed to meet a variety of quality control demands.
Brand: Thermo Scientific™ ESS750
15273.44 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.
- Broad relative humidity range to cover many applications, including humidity levels below ambient.
- Environmentally friendly water-efficient humidification system.
- Minimal condensate in standard applications.
- Integrated water tank; fixed water connection for in-house purified water option available.
- Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
- Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
- Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
- Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
- Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
- Simplified setup with no technician required; install using standard power cord.
- IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
- Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
- Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
- Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Specifications
Audible and Visible | |
66.1 lbs | |
UL, CE | |
750 L | |
Powder-coated Cold Rolled Steel | |
Nema 5-20P | |
0 to 70°C | |
7 in. touchscreen user interface | |
10 - 90% RH | |
4 | |
Temperature and humidity ramping | |
0.46 m² | |
Microprocessor - programmable | |
≤ ±0.1°C @ 20°C to 37°C | |
24 months | |
0.05 - 1 MΩ | |
33.07 in. | |
1080 mm | |
795 mm | |
57.09 in | |
1450 mm | |
24.8 in. | |
630 mm | |
381 kg | |
100/115/220/230 V |
26.5 cu. ft. | |
30 kg | |
USB, Ethernet | |
Powder-coated Cold Rolled Steel | |
3 | |
10 to 90% | |
Humidity Chamber | |
Solid outer door with inner glass door | |
0.1% RHNo | |
2 access ports | |
4.9 ft² | |
Stainless Steel (304/1.4301) | |
0.1°C | |
≤ ±0.5°C @ 20°C to 37°C | |
0.15 to 1 L per day | |
42.52 in. | |
22.6 in. | |
840 mm | |
72.24 in. | |
1835 mm | |
34.84 in. | |
885 mm | |
840 lbs | |
100 - 240V, 50-60Hz, 1400W | |
1400 W |