missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herbimycin A, Tocris Bioscience™
Click to view available options
Quantity:
100 μg
Description
Herbimycin A is an ansamycin antibiotic that acts as a Src family kinase inhibitor. Binds to the SH domain and inhibits the activity of p60v-src and p210BCR-ABL. Exhibits antiangiogenic activity in endothelial cells in vitro. Also inhibits Hsp90 and impairs recovery from heat shock.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Type | Herbimycin A |
CAS | 70563-58-5 |
Quantity | 100 μg |
Molecular Formula | C30H42N2O9 |
Synonym | (15R)-17-demethoxy-15-methoxy-11-O-methyl-geldanamycin |
InChI Key | MCAHMSDENAOJFZ-HYKLGOSOSA-N |
SMILES | CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C=C(C1OC)C2=O)C)OC)OC(=O)N)C)C)OC)OC |
IUPAC Name | [(2S,3R,6S,7R,8E,10R,12E,14E)-2,5,6,11-tetramethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate |
Molecular Weight (g/mol) | 574.671 |
PubChem CID | 133268477 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Herbimycin A, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction