missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22666-1-AP-20UL
This item is not returnable.
View return policy
Description
Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.Specifications
HES5 | |
Polyclonal | |
Unconjugated | |
HES5 | |
bHLHb38, Hairy and enhancer of split 5, HES5, Transcription factor HES 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15208, 388585, 79225 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70120, Q03062, Q5TA89 | |
HES5 | |
HES5 Fusion Protein Ag18270 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |