missing translation for 'onlineSavingsMsg'
Learn More

Hesperadin hydrochloride, Tocris Bioscience™

Product Code. 18054692
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent Aurora kinase B inhibitor

ATP-competitive inhibitor of Aurora B kinase (IC50 = 250 nM). Prevents chromosome alignment and segregation; also induces polyploidy and prevents histone H3-Ser10 phosphorylation. Overrides the spindle assembly checkpoint and induces mitotic exit in monastrol - and taxol -treated HeLa cells.

Chemical Identifiers

CAS 422513-13-1
Molecular Formula C29H32N4O3S
Molecular Weight (g/mol) 516.66
InChI Key GLDSKRNGVVYJAB-DQSJHHFOSA-N
Synonym hesperadin, hesperadine, unii-ptr491os14, z-n-2-oxo-3-phenyl 4-piperidin-1-ylmethyl phenyl amino methylene indolin-5-yl ethanesulfonamide, n-3z-2-oxo-3-phenyl 4-piperidin-1-ylmethyl phenyl amino methylidene-2,3-dihydro-1h-indol-5-yl ethanesulfonamide, n-3z-2-oxo-3-phenyl 4-piperidin-1-ylmethyl phenyl amino methylidene-1h-indol-5-yl ethanesulfonamide, n-2,3-dihydro-2-oxo-3-3z-phenyl 4-1-piperidinylmethyl phenyl amino methylene-1h-indol-5-yl ethanesulfonamide, hesperadin/, d05wsm, ethanesulfonamide, n-2,3-dihydro-2-oxo-3-3z-phenyl 4-1-piperidinylmethyl phenyl amino methylene-1h-indol-5-yl
PubChem CID 10142586
ChEBI CHEBI:70726
IUPAC Name N-[(3Z)-2-oxo-3-[phenyl-[4-(piperidin-1-ylmethyl)anilino]methylidene]-1H-indol-5-yl]ethanesulfonamide
SMILES CCS(=O)(=O)NC1=CC2=C(C=C1)NC(=O)C2=C(C3=CC=CC=C3)NC4=CC=C(C=C4)CN5CCCCC5

Specifications

Quantity 10 mg
Formula Weight 553.12
Percent Purity >99%
Chemical Name or Material Hesperadin hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.