missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Description
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.
Solubility
Soluble in ammonia and water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.
Solubility
Soluble in ammonia and water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Density | 1.71 |
Odor | Odorless |
Linear Formula | [Co(NH3)6]Cl3 |
Quantity | 25 g |
Merck Index | 14,4675 |
Solubility Information | Soluble in ammonia and water. |
Formula Weight | 267.48 |
Percent Purity | 99% |
Chemical Name or Material | Hexaamminecobalt(III) chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'