missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminecobalt(III) chloride, 99%

Product Code. 11492837
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11492837

Brand: Thermo Scientific Alfa Aesar A15470.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10534-89-1 | H18Cl3CoN6

Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaamminecobalt(III) chloride is a component of structural biology methods, especially for DNA or RNA, where the positive ions stabilize the tertiary structure of the phosphate backbone, which favors to solve their structure by X-ray crystallography. It serves as an excellent antibacterial and antiviral property.

Solubility
Soluble in ammonia and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10534-89-1
Molecular Formula H18Cl3CoN6
MDL Number MFCD00036304

Specifications

Density 1.71
Odor Odorless
Linear Formula [Co(NH3)6]Cl3
Quantity 100 g
Merck Index 14,4675
Solubility Information Soluble in ammonia and water.
Formula Weight 267.48
Percent Purity 99%
Chemical Name or Material Hexaamminecobalt(III) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.