Learn More
Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
£13.57 - £110.55
Chemical Identifiers
CAS | 10534-89-1 |
---|---|
Molecular Formula | H18Cl3CoN6 |
Molecular Weight (g/mol) | 267.47 |
MDL Number | MFCD00036304 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10782582
|
Thermo Scientific Acros
405310050 |
5 g | Glass bottle |
£13.57
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10442761
|
Thermo Scientific Acros
405310250 |
25 g | Glass bottle |
£35.98
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422975
|
Thermo Scientific Acros
405311000 |
100 g | Glass bottle |
£110.55
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10534-89-1 | |
H18Cl3CoN6 | |
267.47 | |
MFCD00036304 |
Specifications
217°C | |
Powder | |
H18Cl3CoN6 | |
MFCD00036304 | |
Solubility in water: soluble | |
267.47 | |
Hexaamminecobalt(III) chloride |
10534-89-1 | |
98.5% min. (Argentometry) | |
[Co(NH3)6]Cl3 | |
15, 4710 | |
267.47 | |
99% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is di
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 234-103-9
- Hexaamminecobalt(III) chloride
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H351-Suspected of causing cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Hexaamminecobalt(III) chloride
RUO – Research Use Only