Learn More
Hexacarbonylchromium, 99%, Thermo Scientific Chemicals
£35.48 - £457.28
Chemical Identifiers
CAS | 13007-92-6 |
---|---|
Molecular Formula | C6CrO6 |
Molecular Weight (g/mol) | 220.056 |
MDL Number | MFCD00010945 |
InChI Key | KOTQLLUQLXWWDK-UHFFFAOYSA-N |
Synonym | chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10gShow More |
PubChem CID | 25589 |
ChEBI | CHEBI:33031 |
IUPAC Name | carbon monoxide;chromium |
SMILES | [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382148
|
Thermo Scientific Alfa Aesar
013061.03 |
1 g |
£35.48
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392148
|
Thermo Scientific Alfa Aesar
013061.09 |
10 g |
£109.55
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372148
|
Thermo Scientific Alfa Aesar
013061.18 |
50 g |
£457.28
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hexacarbonylchromium is widely used as a catalyst for isomerization of alkenes. It is involved in the hydrogenation of 1,3-conjugated dienes and oxidation of allylic and benzylic positions. It acts as a precursor to prepare phenanthrenes, tricarbonyl(arene)chromium and Fischer carbine complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13007-92-6 | |
220.056 | |
KOTQLLUQLXWWDK-UHFFFAOYSA-N | |
25589 | |
carbon monoxide;chromium |
C6CrO6 | |
MFCD00010945 | |
chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10gShow More | |
CHEBI:33031 | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr] |
Specifications
∽152°C (decomposition) | |
White | |
99% | |
MFCD00010945 | |
Soluble in ether,chloroform,tetrahydropyran and methylene chloride. Insoluble in water. | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr] | |
220.056 | |
CHEBI:33031 | |
Light sensitive | |
1.77 |
13007-92-6 | |
Crystals | |
C6CrO6 | |
chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g | |
KOTQLLUQLXWWDK-UHFFFAOYSA-N | |
carbon monoxide;chromium | |
25589 | |
220.06 | |
0.35 hPa | |
Hexacarbonylchromium |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P333+P313-P363-P501c
H302+H312+H332-H317-H350i
missing translation for 'dotInformation' : DOT Class: 6.1, Packing Group: III
missing translation for 'einecsNumber' : 235-852-4
missing translation for 'rtecsNumber' : GB5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.