missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorobenzene, 99%, Thermo Scientific Chemicals
£30.50 - £559.00
Chemical Identifiers
CAS | 392-56-3 |
---|---|
Molecular Formula | C6F6 |
Molecular Weight (g/mol) | 186.06 |
MDL Number | MFCD00000288 |
InChI Key | ZQBFAOFFOQMSGJ-UHFFFAOYSA-N |
Synonym | hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus |
PubChem CID | 9805 |
ChEBI | CHEBI:38589 |
IUPAC Name | 1,2,3,4,5,6-hexafluorobenzene |
SMILES | C1(=C(C(=C(C(=C1F)F)F)F)F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10000930
|
Thermo Scientific Acros
120540100 |
10 mL | Glass bottle |
£30.50
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209503
|
Thermo Scientific Acros
120541000 |
100 mL | Glass bottle |
£161.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15129163
|
Thermo Scientific Acros
120545000 |
500 mL | Glass Bottle |
£559.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
392-56-3 | |
186.06 | |
ZQBFAOFFOQMSGJ-UHFFFAOYSA-N | |
9805 | |
1,2,3,4,5,6-hexafluorobenzene |
C6F6 | |
MFCD00000288 | |
hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus | |
CHEBI:38589 | |
C1(=C(C(=C(C(=C1F)F)F)F)F)F |
Specifications
392-56-3 | |
Colorless to Yellow | |
81.0°C to 82.0°C (743.0 mmHg) | |
Authentic | |
C6F6 | |
1.3760 to 1.3780 | |
15,4722 | |
hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus | |
ZQBFAOFFOQMSGJ-UHFFFAOYSA-N | |
1,2,3,4,5,6-hexafluorobenzene | |
9805 | |
186.06 | |
Liquid |
3.7°C to 4.1°C | |
1.6120g/mL | |
10°C | |
98.5% min. (GC) | |
C6F6 | |
MFCD00000288 | |
1.612 | |
Solubility in water: immiscible. Other solubilities: soluble in ethanol | |
C1(=C(C(=C(C(=C1F)F)F)F)F)F | |
186.06 | |
CHEBI:38589 | |
99% | |
Hexafluorobenzene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-876-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- benzene fluoro 6
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only