missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorosilicic acid, 35% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.
Solubility
Miscible with water.
Notes
Incompatible in strong oxidizing agents, metals, alkalis, strong acids, stoneware and glass.
Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.
Solubility
Miscible with water.
Notes
Incompatible in strong oxidizing agents, metals, alkalis, strong acids, stoneware and glass.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 108°C to 109°C |
Quantity | 100 g |
Linear Formula | H2SiF6 |
UN Number | UN1778 |
Merck Index | 14,4182 |
Formula Weight | 144.09 |
Concentration or Composition (by Analyte or Components) | 35% w/w aq. soln. |
Chemical Name or Material | Hexafluorosilicic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'