missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Specifications
Density | 1.8000g/mL |
Packaging | Glass bottle |
Refractive Index | 1.3490 to 1.3570 |
Quantity | 250 mg |
Specific Gravity | 1.8 |
Synonym | ultramark1621, ultramark 1621, hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine |
Formula Weight | 2120 |
Chemical Name or Material | Hexakis(1H, 1H, ωH-perfluoroalkoxy)phosphazene |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'