missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylbenzene, 99+%

Product Code. 11458890
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11458890

Brand: Thermo Scientific Alfa Aesar A10294.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-85-4 | C12H18 | 162.276 g/mol

Hexamethylbenzene is used as pharmaceutical intermediates, in organic synthesis of compounds, as a solvent for He-NMR spectroscopy. Used as a ligand in organometallic chemistry. Reaction with dimethyldioxirane, gives the major product, an unusual oxepane triepoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylbenzene is used as pharmaceutical intermediates, in organic synthesis of compounds, as a solvent for He-NMR spectroscopy. Used as a ligand in organometallic chemistry. Reaction with dimethyldioxirane, gives the major product, an unusual oxepane triepoxide.

Solubility
Insoluble in water

Notes
Incompatible with nitromethane, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-85-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008523
InChI Key YUWFEBAXEOLKSG-UHFFFAOYSA-N
Synonym hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene;
PubChem CID 6908
ChEBI CHEBI:39001
IUPAC Name 1,2,3,4,5,6-hexamethylbenzene
SMILES CC1=C(C(=C(C(=C1C)C)C)C)C

Specifications

Melting Point 163°C to 167°C
Density 1.063
Boiling Point 265°C
Quantity 25 g
Beilstein 1905834
Solubility Information Insoluble in water
Formula Weight 162.28
Percent Purity ≥99%
Chemical Name or Material Hexamethylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.