missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylcyclotrisiloxane, 97%, Thermo Scientific Chemicals

Product Code. 11416216
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible in strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-05-9
Molecular Formula C6H18O3Si3
Molecular Weight (g/mol) 222.46
MDL Number MFCD00005943
InChI Key HTDJPCNNEPUOOQ-UHFFFAOYSA-N
Synonym hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185
PubChem CID 10914
IUPAC Name 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Boiling Point 134°C
Melting Point 64°C to 66°C
Quantity 100 g
UN Number UN1325
Beilstein 1766829
Flash Point 35°C (95°F)
Solubility Information Slightly soluble in water.
Formula Weight 222.47
Percent Purity 97%
Sensitivity Moisture sensitive
Chemical Name or Material Hexamethylcyclotrisiloxane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.