Learn More
Hexamethyldisilane, 98+%, Thermo Scientific Chemicals
£14.10 - £111.56
Chemical Identifiers
CAS | 1450-14-2 |
---|---|
Molecular Formula | C6H18Si2 |
Molecular Weight (g/mol) | 146.38 |
MDL Number | MFCD00008258 |
InChI Key | NEXSMEBSBIABKL-UHFFFAOYSA-N |
Synonym | hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane |
PubChem CID | 74057 |
IUPAC Name | trimethyl(trimethylsilyl)silane |
SMILES | C[Si](C)(C)[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464534
|
Thermo Scientific Alfa Aesar
A13155.06 |
5 g | N/A | ||||||
11474534
|
Thermo Scientific Alfa Aesar
A13155.14 |
25 g |
£43.82
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484534
|
Thermo Scientific Alfa Aesar
A13155.22 |
100 g |
£111.56
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.
Solubility
Insoluble in water. Soluble in alcohol, ether and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
Chemical Identifiers
1450-14-2 | |
146.38 | |
NEXSMEBSBIABKL-UHFFFAOYSA-N | |
74057 | |
C[Si](C)(C)[Si](C)(C)C |
C6H18Si2 | |
MFCD00008258 | |
hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane | |
trimethyl(trimethylsilyl)silane |
Specifications
112°C to 113°C | |
1450-14-2 | |
MFCD00008258 | |
1633463 | |
hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane | |
C[Si](C)(C)[Si](C)(C)C | |
146.38 | |
146.38 | |
Odorless |
12°C to 13°C | |
C6H18Si2 | |
UN1993 | |
−1°C (30°F) | |
NEXSMEBSBIABKL-UHFFFAOYSA-N | |
trimethyl(trimethylsilyl)silane | |
74057 | |
≥98% | |
Hexamethyldisilane |
Safety and Handling
GHS H Statement
H225-H319-H317-H335
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'rtecsNumber' : JM9170000
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.