missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilazane, 98+%, Thermo Scientific Chemicals

£28.34 - £307.53

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.395
MDL Number MFCD00008259
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11492108
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A15139.AE
100 mL
£28.34
100mL
Please to purchase this item. Need a web account? Register with us today!
11402118
missing translation for 'viewDocuments' Détails de la promotion
Thermo Scientific Alfa Aesar
A15139.AP
500 mL
£81.81
500mL
Please to purchase this item. Need a web account? Register with us today!
11412118
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A15139.0F
2500 mL
£307.53
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

Solubility
Miscible with methanol, chloroform, ethyl ether and benzene. Immiscible with water.                    

Notes
Incompatible with strong oxidizing agents and strong acids.  
Spécification

Identifiants chimiques

999-97-3
161.395
FFUAGWLWBBFQJT-UHFFFAOYSA-N
13838
[dimethyl-(trimethylsilylamino)silyl]methane
C6H19NSi2
MFCD00008259
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
CHEBI:85068
C[Si](C)(C)N[Si](C)(C)C

Spécification

999-97-3
0.774
8°C (46°F)
Odorless
(CH3)3SiNHSi(CH3)3
MFCD00008259
635752
14,4689
Miscible with methanol,chloroform,ethyl ether and benzene. Immiscible with water.
C[Si](C)(C)N[Si](C)(C)C
161.395
CHEBI:85068
≥98%
Hexamethyldisilazane
-78°C
126°C
98+%
C6H19NSi2
1.408
UN3286
Moisture sensitive
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
FFUAGWLWBBFQJT-UHFFFAOYSA-N
[dimethyl-(trimethylsilylamino)silyl]methane
13838
161.4
Liquid
Vidéos
Sécurité et traitement

Sécurité et traitement

GHS H Statement
H225-H311-H331-H314-H318-H302
Highly flammable liquid and vapour.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H311-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 213-668-5

missing translation for 'rtecsNumber' : JM9230000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Moisture sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

Identifiant produit
  • Hexamethyldisilazane

Mention d'avertissement
  • Danger

Catégorie de danger
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2

Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Informations supplémentaires
  • MIXTURE LIST-Contains : Hexamethyldisilazane

FDS
Documents

Documents

Certificats

RUO – Research Use Only