missing translation for 'onlineSavingsMsg'
Learn More
Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific Chemicals Product Code.: 11319215

Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific Chemicals

Product Code. 11319215
100 mL
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11319215

Brand: Thermo Scientific Alfa Aesar L16519.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

Solubility
Miscible with methanol, ethyl ether, chloroform and benzene. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
MDL Number MFCD00008259
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethylShow MoreShow Less
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Boiling Point 126°C
Melting Point -78°C
Quantity 100 mL
Linear Formula (CH3)3SiNHSi(CH3)3
UN Number UN3286
Beilstein 635752
Flash Point 8°C (46°F)
Merck Index 14,4689
Solubility Information Miscible with methanol,ethyl ether,chloroform and benzene. Immiscible with water.
Molecular Weight (g/mol) 161.395
Formula Weight 161.4
Percent Purity ≥99%
Grade Electronic
Odor Odorless
Refractive Index 1.408
Sensitivity Moisture sensitive
Density 0.774
Chemical Name or Material Hexamethyldisilazane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN20L004 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberL16519.AP,L16519.AC,L16519.AE
Lot NumberZ26K0431 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog NumberL16519.AP,L16519.AC,L16519.AE
Lot NumberW18K034 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberL16519.AP,L16519.AC,L16519.AE
Lot NumberQ09K038 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog NumberL16519.AP,L16519.AC,L16519.AE
Lot NumberZ08H038 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL16519.AP,L16519.AC,L16519.AE
compliance-icons compliance-icons
Product Identifier
  • Hexamethyldisilazane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Hexamethyldisilazane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.