missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethyldisilazane, Honeywell™
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 999-97-3 |
Physical Form | Liquid |
Quantity | 1 L |
Flash Point | 11 °C |
PubChem CID | 13838 |
ChEBI | CHEBI:85068 |
Percent Purity | ≥98% |
Grade | Semiconductor |
Packaging | Plastic Bottle |
Refractive Index | 1.4070 - 1.4090 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'