missing translation for 'onlineSavingsMsg'
Learn More
Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals Product Code.: 11443814

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Product Code. 11443814
2500 mL
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11443814

Brand: Thermo Scientific Alfa Aesar A11848.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-46-0
Molecular Formula C6H18OSi2
MDL Number MFCD00008265
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxideShow MoreShow Less
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Melting Point -67°C
Density 0.764
Boiling Point 100°C to 101°C
Flash Point −2°C (28°F)
Odor Characteristic
Linear Formula (CH3)3SiOSi(CH3)3
Refractive Index 1.377
Quantity 2500 mL
UN Number UN1993
Beilstein 1736258
Solubility Information Slightly miscible with water.
Molecular Weight (g/mol) 162.379
Formula Weight 162.38
Percent Purity ≥98%
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10217352 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberA11848.AP,A11848.AE,A11848.0F
Lot Number10244511 Certificate TypeCertificate of Analysis Date10/11/2022 Catalog NumberA11848.AP,A11848.AE,A11848.0F
Lot NumberW30A060 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11848.AP,A11848.AE,A11848.0F
Lot Number10168305 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11848.AP,A11848.AE,A11848.0F
Lot Number10206131 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11848.AP,A11848.AE,A11848.0F
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Hexamethyldisiloxane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.