missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylenetetramine, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
CAS | 100-97-0 |
Flash Point | 250.00°C |
Packaging | Poly Bottle |
Molecular Formula | C6H12N4 |
Quantity | 500 g |
Specific Gravity | 1L = 1.33kg |
Synonym | methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine |
InChI Key | VKYKSIONXSXAKP-UHFFFAOYSA-N |
SMILES | C1N2CN3CN1CN(C2)C3 |
Molecular Weight (g/mol) | 140.19 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'