missing translation for 'onlineSavingsMsg'
Learn More
Riedel-de Haën Hexane CHROMASOLV™ Product Code.: 16653954

Riedel-de Haën Hexane CHROMASOLV™

Product Code. 16653954
200 L
Click to view available options
Quantity:
7 L
18 L
45 L
200 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 16653954

Brand: Riedel-de Haën 34859200L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Hexane CHROMASOLV™ for HPLC and organic synthesis, ≥97.0% (GC).

  • For HPLC and organic synthesis.
  • High-purity, multipurpose solvents.
  • High lot-to-lot consistency.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-Hexane
IUPAC Name hexane
SMILES CCCCCC

Specifications

pH Not applicable
Melting Point -95°C
Color Colorless
CAS Max % 100.00%
Density 0.660 to 0.668 g/cm3 (20°C)
Boiling Point 66°C to 69°C (1.013 hPa)
Quantity 200 L
Flash Point 7.4 %(V)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line CHROMASOLV™
Linear Formula CH3(CH2)4CH3
Vapor Pressure 160 hPa (20°C)
UN Number UN1208
Viscosity 0.31 mPa.s (20°C)
Beilstein 1730733
Concentration 100.00%
Percent Purity ≥97.00%
Grade HPLC
Chemical Name or Material Hexane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Riedel-de Haën Hexane CHROMASOLV™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.