missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, CHROMASOLV™, ≥97%, LC-MS, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS
£107.00 - £291.92
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15614760
|
Honeywell-Chromasolv
34986-1L |
1 L | N/A | ||||||
15624760
|
Honeywell-Chromasolv
34986-2.5L |
2.5 L |
£291.92
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanesShow More | |
CHEBI:29021 | |
CCCCCC |
Specifications
110-54-3 | |
0.077 | |
453°F | |
CH3(CH2)4CH3 | |
5.2 psi (37.7°C), 132 mmHg (20°C), 256 mmHg (37.7°C) | |
1730733 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18g/mol | |
LC-MS CHROMASOLV™ |
0.659g/mL at 25°C | |
−26°C (−14.8°F) | |
C6H14 | |
MFCD02179311 | |
UN1208 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥97% | |
Hexane |
Safety and Handling
P201-P210-P273-P301 + P310-P308 + P313-P331
missing translation for 'rtecsNumber' : MN9275000
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hexane, CHROMASOLV™, ≥97%, LC-MS, Honeywell Riedel-de Haën™