missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane Fraction, Specified Reagent for General Laboratory Work, SLR, Fisher Chemical™
£33.70 - £697.00
Chemical Identifiers
CAS | 92112-69-1 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD00009520 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10549380
|
Fisher Chemical
H/0350/08 |
500 mL | Amber Glass Bottle |
£33.70
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10655682
|
Fisher Chemical
H/0350/15 |
1 L | Amber Glass Bottle |
£45.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10735141
|
Fisher Chemical
H/0350/17 |
2.5 L | Amber Glass Bottle |
£104.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10753124
|
Fisher Chemical
H/0350/PB17 |
2.5 L | Plastic-coated Glass Bottle |
£132.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10407070
|
Fisher Chemical
H/0350/21 |
5 L | Metal Can |
£135.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10369899
|
Fisher Chemical
H/0350/FP21 |
5 L | Plastic Container |
£126.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10050190
|
Fisher Chemical
H/0350/25 |
25 L | Metal Can |
£697.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
92112-69-1 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD00009520 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
Colorless | |
Liquid | |
MFCD00009520 | |
0.31 mPaS at 20°C | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18g/mol |
92112-69-1 | |
69°C | |
C6H14 | |
160mbar at 20°C | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CCCCCC | |
86.18 | |
CHEBI:29021 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hexane fraction from petroleum
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.