missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, Laboratory Reagent, ≥95%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
20 L
25 L
4 x 2.5 L
4 x 5 L
6 x 1 L
190 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Poly Pail
Stainless Steel Can
Unit Size:
1 L
10 L
190 L
2.5 L
20 L
25 L
5 L
500mL
Pack of 4
Pack of 6
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | -95°C |
| Color | Colorless |
| Density | 0.659g/mL at 25°C |
| Boiling Point | 69°C |
| Quantity | 5 L |
| Explosion Limit | 0.077 |
| Flash Point | −26°C (−14.8°F) |
| Ignition Point | 453°F |
| Packaging | Aluminum Bottle |
| Physical Form | Liquid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction