missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Laboratory Reagent, ≥95%, Honeywell™

Product Code. 15631860 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
20 L
25 L
4 x 2.5 L
4 x 5 L
6 x 1 L
190 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Poly Pail
Stainless Steel Can
Unit Size:
1 L
10 L
190 L
2.5 L
20 L
25 L
5 L
500mL
Pack of 4
Pack of 6
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15631860

Brand: Honeywell Research Chemicals 208752190L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Laboratory Reagent,≥95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.659g/mL at 25°C
Boiling Point 69°C
Quantity 190 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Packaging Metal Drum
Physical Form Liquid
Refractive Index n20/D 1.375(lit.)
Linear Formula CH3(CH2)4CH3
Vapor Pressure 132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C)
UN Number UN1208
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥95%
Grade Laboratory Reagent
Chemical Name or Material Hexane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.