missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, Mixture of Isomers, ACS Reagent, Honeywell Riedel-de Haen™
Click to view available options
500 mL
Specifications
Melting Point | -154°C |
Color | Colorless |
UN Number | UN1208 |
Boiling Point | 48°C to 65°C (1013 hPa) |
Quantity | 500 mL |
Chemical Name or Material | Hexane, mixture of isomers |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'