missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, mixture of isomers, CHROMASOLV™, for HPLC, ≥98.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Melting Point | -95°C (literature) |
Absorbance | max. 0.01 at 250 nm, max. 0.02 at 240 nm, max. 0.05 at 230 nm, max. 0.10 at 220 nm, max. 0.30 at 210 nm, max. 1.0 at 195 nm, max. 0.01 at 400 nm |
CAS | 110-54-3 |
Density | 0.672 g/mL at 25 °C(lit.) |
Boiling Point | 48°C to 65°C |
Quantity | 1 L |
Flash Point | −26°C |
Packaging | Glass Bottle |
Molecular Formula | C6H14 |
Linear Formula | C6H14 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'