missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, ReagentPlus™, ≥99%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.659g/mL at 25°C |
Boiling Point | 66°C to 69°C |
Quantity | 6 x 500 mL |
Explosion Limit | 0.077 |
Flash Point | −26°C (−14.8°F) |
Ignition Point | 453°F |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
Safety and Handling




Product Identifier
- Hexane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : n-hexane
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hexane, ReagentPlus™, ≥99%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction