missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ≤97%, Extra Dry, mixture of isomers, AcroSeal™, Thermo Scientific Chemicals

£25.13 - £72.26

Specifications

Chemical Name or Material Hexanes
CAS 92112-69-1, 110-54-3
Density 0.6590g/mL
Identification Pass Test
Packaging AcroSeal™ Glass Bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10401674
View Documents
Thermo Scientific Acros
610371000
100 mL AcroSeal™ Glass Bottle
£25.13
100mL
Please to purchase this item. Need a web account? Register with us today!
10431484
View Documents
Thermo Scientific Acros
610370010
1 L AcroSeal™ Glass Bottle
£72.26
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexanes
0.6590g/mL
AcroSeal™ Glass Bottle
CH3(CH2)4CH3
Vapor Pressure: 160mbar at 20°C
50ppm max.
Hex
≤97%
92112-69-1, 110-54-3
Pass Test
C6H14
MFCD00009520
01, 142
14, 4694
86.18
Extra Dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 203-777-6

missing translation for 'rtecsNumber' : MN9275000

missing translation for 'tsca' : TSCA

Product Identifier
  • Hexanes

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.