missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 411540100

Code : 29

Additional Details : CAS Number : 92112-69-1 Weight : 6.59000kg Transport : UN number : 1208 Chem class : 3 Pack group : II

 View more versions of this product

Product Code. 10073443

  • £175.00 / 10 L
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexanes
Mixture of Isomers
Colorless
69.0°C
−22°C
Plastic Drum
1.3748 to 1.3810
0.659
CH3(CH2)4CH3
Vapor Pressure: 160mbar at 20°C
01, 142
Hex
86.18
Pure
-95.0°C
92112-69-1,110-54-3
0.6590g/mL
10 L
Authentic
Liquid
99%
C6H14
MFCD00009520
0.31 mPa.s (20°C)
14, 4694
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only