missing translation for 'onlineSavingsMsg'
Learn More

Hexanes (Certified ACS), Fisher Chemical™

Product Code. 15608755
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass Bottle
Steel Pail

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Color Colorless
Boiling Point 65.5°C
Quantity 20 L
Flash Point -6.7°C (+20°F)
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Assay Percent Range ≥98.5 %
Residue after Evaporation 0.001% max.
Specific Gravity Less-than water
UN Number 1208
Merck Index 15, 4732
Solubility Information Negligible
Formula Weight 86.18
Percent Purity ≥98.5%
Grade Certified ACS
Chemical Name or Material Hexanes
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexanes
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hexane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'