Learn More
Hexanes, for HPLC, contains ca. 95% n-Hexane, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 268360025
61.58 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
Hexanes | |
(1 cm cell vs HPLC−grade water), 0.005 max. at 250nm, 0.01 max. at 240nm, 0.03 max. at 230nm, 0.07 max. at 220nm, 0.25 max. at 210nm, 0.7 max. at 200nm | |
0.6590g/mL | |
Glass Bottle | |
CH3(CH2)4CH3 | |
Vapor Pressure: 160mbar at 20°C | |
14, 4694 | |
86.18 |
(95% n-Hexane) | |
92112-69-1, 110-54-3 | |
2.5 L | |
C6H14 | |
MFCD00009520 | |
01, 142 | |
Hex | |
HPLC |
Safety and Handling
- Hexanes
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only